accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 957060-90-1

IUPAC Name: [2-(2,2,2-trifluoroethoxy)phenyl]boronic acid
Synonyms: 2-(2,2,2-trifluoroethoxy)phenylboronic acid
Molecular Weight (g/mol): 219.95
Molecular Formula: C8H8BF3O3
InChi Key: IFWDRBWGMUCFMH-UHFFFAOYSA-N
SMILES: OB(O)C1=CC=CC=C1OCC(F)(F)F

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results