accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

BOOH3CCH3H3CH3CNOOCH3

CAS RN 957062-84-9

IUPAC Name: 4,4,5,5-tetramethyl-2-(2-methyl-5-nitrophenyl)-1,3,2-dioxaborolane
Synonyms: 4,4,5,5-tetramethyl-2-(2-methyl-5-nitrophenyl)-1,3,2-dioxaborolane
Molecular Weight (g/mol): 263.1
Molecular Formula: C13H18BNO4
InChi Key: LYFUUPOOXAXEEP-UHFFFAOYSA-N
SMILES: CC1=C(C=C(C=C1)[N+]([O-])=O)B1OC(C)(C)C(C)(C)O1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results