accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

BSOONHFHOOH

CAS RN 957121-13-0

IUPAC Name: {4-[(4-fluorophenyl)sulfamoyl]phenyl}boronic acid
Synonyms: 4-[(4-fluorophenyl)sulfamoyl]phenylboronic acid
Molecular Weight (g/mol): 295.09
Molecular Formula: C12H11BFNO4S
InChi Key: MFWQIJUPTCPQPU-UHFFFAOYSA-N
SMILES: OB(O)C1=CC=C(C=C1)S(=O)(=O)NC1=CC=C(F)C=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results