accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

H3COFH3C

CAS RN 95729-22-9

IUPAC Name: 4-fluoro-2-methoxy-1-methylbenzene
Synonyms: 4-fluoro-2-methoxy-1-methylbenzene
Molecular Weight (g/mol): 140.16
Molecular Formula: C8H9FO
InChi Key: CXTIUQIZWCABPV-UHFFFAOYSA-N
SMILES: COC1=CC(F)=CC=C1C

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results