accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 957311-13-6

CH3H3CCH3OONOHOFFF

CAS RN 957311-13-6

IUPAC Name: 1-[(tert-butoxy)carbonyl]-4-{[2-(trifluoromethyl)phenyl]methyl}pyrrolidine-2-carboxylic acid
Synonyms: 1-(tert-butoxycarbonyl)-4-{[2-(trifluoromethyl)phenyl]methyl}pyrrolidine-2-carboxylic acid
Molecular Weight (g/mol): 373.37
Molecular Formula: C18H22F3NO4
InChi Key: LXKNKSPHSOFZBM-UHFFFAOYNA-N
SMILES: CC(C)(C)OC(=O)N1CC(CC2=CC=CC=C2C(F)(F)F)CC1C(O)=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results