accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

H3CHNO

CAS RN 958443-30-6

IUPAC Name: methyl[(4-phenyloxan-4-yl)methyl]amine
Synonyms: methyl[(4-phenyloxan-4-yl)methyl]amine
Molecular Weight (g/mol): 205.3
Molecular Formula: C13H19NO
InChi Key: OHCOJCIAAGLEHJ-UHFFFAOYSA-N
SMILES: CNCC1(CCOCC1)C1=CC=CC=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results