accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 958443-39-5

IUPAC Name: 4-methyl-2-(pyridin-3-yl)-1,3-thiazole-5-carbaldehyde
Synonyms: 4-methyl-2-(pyridin-3-yl)-1,3-thiazole-5-carbaldehyde
Molecular Weight (g/mol): 204.25
Molecular Formula: C10H8N2OS
InChi Key: KHHZRQVJLBGSNM-UHFFFAOYSA-N
SMILES: CC1=C(SC(=N1)C1=CN=CC=C1)C=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results