accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 959705-64-7

IUPAC Name: 12-methoxy-5,7-dimethyl-3-propyl-2,4,8,13-tetraazatricyclo[7.4.0.0²,⁶]trideca-1(13),3,5,7,9,11-hexaene
Synonyms: 12-methoxy-5,7-dimethyl-3-propyl-2,4,8,13-tetraazatricyclo[7.4.0.0²,⁶]trideca-1(13),3,5,7,9,11-hexaene
Molecular Weight (g/mol): 270.34
Molecular Formula: C15H18N4O
InChi Key: YNADXFWEXJTQSZ-UHFFFAOYSA-N
SMILES: CCCC1=NC(C)=C2N1C1=NC(OC)=CC=C1N=C2C

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results