accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 959755-46-5

IUPAC Name: 5,8-dibromo-[1,2,4]triazolo[1,5-a]pyrazine
Synonyms: 5,8-dibromo-[1,2,4]triazolo[1,5-a]pyrazine
Molecular Weight (g/mol): 277.91
Molecular Formula: C5H2Br2N4
InChi Key: IDWOTLNDEITXRA-UHFFFAOYSA-N
SMILES: BrC1=CN=C(Br)C2=NC=NN12

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results