accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 96-05-9

H3CCH2OOCH2

CAS RN 96-05-9

IUPAC Name: prop-2-en-1-yl 2-methylprop-2-enoate
Synonyms: allyl methacrylate
Molecular Weight (g/mol): 126.16
Molecular Formula: C7H10O2
InChi Key: FBCQUCJYYPMKRO-UHFFFAOYSA-N
SMILES: CC(=C)C(=O)OCC=C

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results