accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 96-10-6

IUPAC Name: chlorodiethylalumane
Synonyms: diethylaluminium chloride
Molecular Weight (g/mol): 120.56
Molecular Formula: C4H10AlCl
InChi Key: YNLAOSYQHBDIKW-UHFFFAOYSA-M
SMILES: CC[Al](Cl)CC
Quantity 
  • (1)
  • (2)
  • (1)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results