accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 96-31-1

IUPAC Name: 1,3-dimethylurea
Synonyms: dimethylurea dimethyl urea dimethylurea, sym
Molecular Weight (g/mol): 88.11
Molecular Formula: C3H8N2O
InChi Key: MGJKQDOBUOMPEZ-UHFFFAOYSA-N
SMILES: CNC(=O)NC
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
Boiling Point 
  • (3)
  • (3)
Physical Form 
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (3)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (3)
  • (3)
  • (3)