accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CH3OOH2C

CAS RN 96-33-3

IUPAC Name: methyl prop-2-enoate
Synonyms: methyl acrylate
Molecular Weight (g/mol): 86.09
Molecular Formula: C4H6O2
InChi Key: BAPJBEWLBFYGME-UHFFFAOYSA-N
SMILES: COC(=O)C=C
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
Boiling Point 
  • (2)
  • (5)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (5)
  • (5)
  • (2)
  • (3)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (2)
  • (5)
  • (5)