accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 96-35-5

CH3OOHO

CAS RN 96-35-5

IUPAC Name: methyl 2-hydroxyacetate
Synonyms: methyl 2-hydroxyacetate
Molecular Weight (g/mol): 90.08
Molecular Formula: C3H6O3
InChi Key: GSJFXBNYJCXDGI-UHFFFAOYSA-N
SMILES: COC(=O)CO
Boiling Point 
  • (3)
  • (2)
Molecular Weight (g/mol) 
  • (3)
  • (2)
Quantity 
  • (2)
  • (2)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (2)
  • (3)
  • (3)
  • (2)
  • (3)
  • (2)
  • (2)
  • (2)
  • (1)
  • (2)