accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 96-45-7

SNHHN

CAS RN 96-45-7

IUPAC Name: imidazolidine-2-thione
Synonyms: 2-imidazolidinethione
Molecular Weight (g/mol): 102.16
Molecular Formula: C3H6N2S
InChi Key: PDQAZBWRQCGBEV-UHFFFAOYSA-N
SMILES: S=C1NCCN1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results