CAS RN 96-47-9
2-Methyltetrahydrofuran, 99+%, Extra Dry, stabilized, AcroSeal™
CAS: 96-47-9 Molecular Formula: C5H10O Molecular Weight (g/mol): 86.13 MDL Number: MFCD00005367 InChI Key: JWUJQDFVADABEY-UHFFFAOYSA-N Synonym: 2-methyltetrahydrofuran,methyltetrahydrofuran,tetrahydrosylvan,tetrahydro-2-methylfuran,furan, tetrahydro-2-methyl,furan, 2-methyl-tetrahydro,2-methyl tetrahydrofuran,2-methyl-tetrahydrofuran,2-methyl-tetrahydro-furan,methyl tetrahydrofuran PubChem CID: 7301 IUPAC Name: 2-methyloxolane SMILES: CC1CCCO1
2-Methyltetrahydrofuran, 99+%, Extra Dry, stabilizer free, AcroSeal™
CAS: 96-47-9 Molecular Formula: C5H10O Molecular Weight (g/mol): 86.13 MDL Number: MFCD00005367 InChI Key: JWUJQDFVADABEY-UHFFFAOYSA-N Synonym: 2-methyltetrahydrofuran,methyltetrahydrofuran,tetrahydrosylvan,tetrahydro-2-methylfuran,furan, tetrahydro-2-methyl,furan, 2-methyl-tetrahydro,2-methyl tetrahydrofuran,2-methyl-tetrahydrofuran,2-methyl-tetrahydro-furan,methyl tetrahydrofuran PubChem CID: 7301 IUPAC Name: 2-methyloxolane SMILES: CC1CCCO1
2-Methyltetrahydrofuran, For Grignard reaction, anhydrous, stabilized
CAS: 96-47-9 Molecular Formula: C5H10O Molecular Weight (g/mol): 86.13 InChI Key: JWUJQDFVADABEY-UHFFFAOYSA-N Synonym: 2-methyltetrahydrofuran,methyltetrahydrofuran,tetrahydrosylvan,tetrahydro-2-methylfuran,furan, tetrahydro-2-methyl,furan, 2-methyl-tetrahydro,2-methyl tetrahydrofuran,2-methyl-tetrahydrofuran,2-methyl-tetrahydro-furan,methyl tetrahydrofuran PubChem CID: 7301 IUPAC Name: 2-methyloxolane SMILES: CC1CCCO1
2-Methyltetrahydrofuran, 99+%, Extra Dry over Molecular Sieve, Stabilized, AcroSeal™
CAS: 96-47-9 Molecular Formula: C5H10O Molecular Weight (g/mol): 86.13 MDL Number: MFCD00005367 InChI Key: JWUJQDFVADABEY-UHFFFAOYSA-N Synonym: 2-methyltetrahydrofuran,methyltetrahydrofuran,tetrahydrosylvan,tetrahydro-2-methylfuran,furan, tetrahydro-2-methyl,furan, 2-methyl-tetrahydro,2-methyl tetrahydrofuran,2-methyl-tetrahydrofuran,2-methyl-tetrahydro-furan,methyl tetrahydrofuran PubChem CID: 7301 IUPAC Name: 2-methyloxolane SMILES: CC1CCCO1
2-Methyltetrahydrofuran, 99+%, pure, stabilized
CAS: 96-47-9 Molecular Formula: C5H10O Molecular Weight (g/mol): 86.13 InChI Key: JWUJQDFVADABEY-UHFFFAOYSA-N Synonym: 2-methyltetrahydrofuran,methyltetrahydrofuran,tetrahydrosylvan,tetrahydro-2-methylfuran,furan, tetrahydro-2-methyl,furan, 2-methyl-tetrahydro,2-methyl tetrahydrofuran,2-methyl-tetrahydrofuran,2-methyl-tetrahydro-furan,methyl tetrahydrofuran PubChem CID: 7301 IUPAC Name: 2-methyloxolane SMILES: CC1CCCO1
2-Methyltetrahydrofuran, 99%, stab. with ca 150-400ppm BHT
CAS: 96-47-9 Molecular Formula: C5H10O Molecular Weight (g/mol): 86.134 MDL Number: MFCD00005367 InChI Key: JWUJQDFVADABEY-UHFFFAOYSA-N Synonym: 2-methyltetrahydrofuran,methyltetrahydrofuran,tetrahydrosylvan,tetrahydro-2-methylfuran,furan, tetrahydro-2-methyl,furan, 2-methyl-tetrahydro,2-methyl tetrahydrofuran,2-methyl-tetrahydrofuran,2-methyl-tetrahydro-furan,methyl tetrahydrofuran PubChem CID: 7301 IUPAC Name: 2-methyloxolane SMILES: CC1CCCO1
2-Methyltetrahydrofuran, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
2-Methyltetrahydrofuran, ReagentPlus™, ≥99.5%,Solstice
CAS: 96-47-9 Molecular Formula: C5H10O Molecular Weight (g/mol): 86.134 MDL Number: MFCD00005367 InChI Key: JWUJQDFVADABEY-UHFFFAOYSA-N Synonym: 2-methyltetrahydrofuran,methyltetrahydrofuran,tetrahydrosylvan,tetrahydro-2-methylfuran,furan, tetrahydro-2-methyl,furan, 2-methyl-tetrahydro,2-methyl tetrahydrofuran,2-methyl-tetrahydrofuran,2-methyl-tetrahydro-furan,methyl tetrahydrofuran PubChem CID: 7301 IUPAC Name: 2-methyloxolane SMILES: CC1CCCO1
2-Methyltetrahydrofuran, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
2-Methyltetrahydrofuran, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.