accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CH3N

CAS RN 96-54-8

IUPAC Name: 1-methyl-1H-pyrrole
Synonyms: N-methylpyrrole
Molecular Weight (g/mol): 81.12
Molecular Formula: C5H7N
InChi Key: OXHNLMTVIGZXSG-UHFFFAOYSA-N
SMILES: CN1C=CC=C1
Molecular Weight (g/mol) 
  • (3)
  • (5)
Quantity 
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (2)
Boiling Point 
  • (2)
  • (3)
Percent Purity 
  • (3)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (3)
  • (3)
  • (3)
  • (5)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (2)
  • (2)
  • (3)
  • (3)
  • (2)
  • (5)