accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 96-76-4

IUPAC Name: 2,4-di-tert-butylphenol
Synonyms: 2,4-di-T-butylphenol
Molecular Weight (g/mol): 206.33
Molecular Formula: C14H22O
InChi Key: ICKWICRCANNIBI-UHFFFAOYSA-N
SMILES: CC(C)(C)C1=CC=C(O)C(=C1)C(C)(C)C
Molecular Weight (g/mol) 
  • (3)
  • (4)
  • (4)
Percent Purity 
  • (2)
  • (3)
  • (6)
Quantity 
  • (2)
  • (1)
  • (1)
  • (2)
  • (1)
  • (1)
  • (2)
  • (1)
Boiling Point 
  • (4)
  • (2)
Physical Form 
  • (6)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (3)
  • (3)
  • (4)
  • (4)
  • (2)
  • (3)
  • (6)
  • (2)
  • (1)
  • (1)
  • (2)
  • (1)
  • (1)
  • (2)
  • (1)
  • (4)
  • (2)
  • (6)