accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 96-98-0

IUPAC Name: 4-methyl-3-nitrobenzoic acid
Synonyms: benzoic acid,4-methyl-3-nitro
Molecular Weight (g/mol): 181.15
Molecular Formula: C8H7NO4
InChi Key: BBEWSMNRCUXQRF-UHFFFAOYSA-N
SMILES: CC1=CC=C(C=C1[N+]([O-])=O)C(O)=O
Molecular Weight (g/mol) 
  • (2)
  • (6)
Quantity 
  • (1)
  • (1)
  • (2)
  • (1)
  • (1)
  • (2)
Physical Form 
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results