accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 96193-27-0

NHN

CAS RN 96193-27-0

IUPAC Name: octahydropyrrolo[1,2-a]pyrazine
Synonyms: octahydropyrrolo[1,2-a]pyrazine
Molecular Weight (g/mol): 126.2
Molecular Formula: C7H14N2
InChi Key: FTTATHOUSOIFOQ-UHFFFAOYNA-N
SMILES: C1CC2CNCCN2C1
Boiling Point 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results