accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 962-58-3

CH3OPOOCH3OCH3NNCH3H3C

CAS RN 962-58-3

IUPAC Name: diethyl 6-methyl-2-(propan-2-yl)pyrimidin-4-yl phosphate
Synonyms: diazinon, O-analog
Molecular Weight (g/mol): 288.28
Molecular Formula: C12H21N2O4P
InChi Key: VBLJFQYCTRKKKF-UHFFFAOYSA-N
SMILES: CCOP(=O)(OCC)OC1=CC(C)=NC(=N1)C(C)C
Molecular Weight (g/mol) 
  • (4)
  • (1)
Quantity 
  • (1)
  • (1)
  • (1)
Solvent or Matrix 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results