accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CH3OHOSBrOBr(Z)

CAS RN 96232-71-2

IUPAC Name: (2Z)-4,5-dibromo-2-[hydroxy(methoxy)methylidene]-2,3-dihydrothiophen-3-one
Synonyms: (2Z)-4,5-dibromo-2-[hydroxy(methoxy)methylidene]thiophen-3-one
Molecular Weight (g/mol): 315.96
Molecular Formula: C6H4Br2O3S
InChi Key: YZPNGCXFJCPJSV-XQRVVYSFSA-N
SMILES: CO\C(O)=C1/SC(Br)=C(Br)C1=O
Quantity 
  • (2)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results