accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CH3NNNH3CCH3

CAS RN 96556-05-7

IUPAC Name: 1,4,7-trimethyl-1,4,7-triazonane
Synonyms: 1,4,7-trimethyl-1,4,7-triazonane
Molecular Weight (g/mol): 171.29
Molecular Formula: C9H21N3
InChi Key: WLDGDTPNAKWAIR-UHFFFAOYSA-N
SMILES: CN1CCN(C)CCN(C)CC1
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (1)
  • (1)
  • (1)