accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 96799-03-0

IUPAC Name: 5-(thiophen-2-yl)-1H-pyrazol-3-amine
Synonyms: 5-(thiophen-2-yl)-1H-pyrazol-3-amine
Molecular Weight (g/mol): 165.21
Molecular Formula: C7H7N3S
InChi Key: TXSOLYKLZBJHFF-UHFFFAOYSA-N
SMILES: NC1=NNC(=C1)C1=CC=CS1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results