CAS RN 96948-64-0
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
2
of
2
results
1
(1RS)-2-[(1,1-Dimethylethyl)amino]-1-(4-hydroxyphenyl)ethanol, Mikromol™
Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.
2
t-Butylnorsynephrine, TRC
CAS: 96948-64-0 Molecular Formula: C12 H19 N O2 Molecular Weight (g/mol): 209.28 Synonym: (1RS)-2-[(1,1-Dimethylethyl)amino]-1-(4-hydroxyphenyl)ethanol,Salbutamol Sulphate Imp. B (EP) IUPAC Name: 4-[2-(tert-butylamino)-1-hydroxyethyl]phenol SMILES: CC(C)(C)NCC(O)c1ccc(O)cc1