accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 97-00-7

IUPAC Name: 1-chloro-2,4-dinitrobenzene
Synonyms: Cdnb DNCB Dncb chlorodinitrobenzene dinitrochlorobenzene
Molecular Weight (g/mol): 202.55
Molecular Formula: C6H3ClN2O4
InChi Key: VYZAHLCBVHPDDF-UHFFFAOYSA-N
SMILES: [O-][N+](=O)C1=CC=C(Cl)C(=C1)[N+]([O-])=O
Quantity 
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
Boiling Point 
  • (3)
  • (3)
Percent Purity 
  • (3)
  • (3)
Physical Form 
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (3)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (3)
  • (3)
  • (3)
  • (3)
  • (3)
  • (1)