accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 97-30-3

IUPAC Name: 2-(hydroxymethyl)-6-methoxyoxane-3,4,5-triol
Synonyms: methyl-α-D-mannoside methyl mannoside mannoside, methyl
Molecular Weight (g/mol): 194.18
Molecular Formula: C7H14O6
InChi Key: HOVAGTYPODGVJG-UHFFFAOYNA-N
SMILES: COC1OC(CO)C(O)C(O)C1O
Molecular Weight (g/mol) 
  • (6)
  • (2)
Quantity 
  • (3)
  • (1)
  • (1)
  • (1)
  • (2)
Percent Purity 
  • (2)
  • (3)
Boiling Point 
  • (3)
Physical Form 
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (1)
  • (6)
  • (2)
  • (3)
  • (1)
  • (1)
  • (1)
  • (2)
  • (2)
  • (3)
  • (3)
  • (3)