accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 97-51-8

IUPAC Name: 2-hydroxy-5-nitrobenzaldehyde
Synonyms: 5-nitrosalicylaldehyde
Molecular Weight (g/mol): 167.12
Molecular Formula: C7H5NO4
InChi Key: IHFRMUGEILMHNU-UHFFFAOYSA-N
SMILES: OC1=CC=C(C=C1C=O)[N+]([O-])=O
Quantity 
  • (2)
  • (2)
  • (3)
  • (1)
Percent Purity 
  • (3)
  • (3)
Physical Form 
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results