accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 97-52-9

IUPAC Name: 2-methoxy-4-nitroaniline
Synonyms: 2-methoxy-4-nitroaniline
Molecular Weight (g/mol): 168.15
Molecular Formula: C7H8N2O3
InChi Key: GVBHRNIWBGTNQA-UHFFFAOYSA-N
SMILES: COC1=CC(=CC=C1N)[N+]([O-])=O
Quantity 
  • (2)
  • (1)
  • (2)
  • (2)
Physical Form 
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results