accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

H3COCH2HO

CAS RN 97-53-0

IUPAC Name: 2-methoxy-4-(prop-2-en-1-yl)phenol
Synonyms: eugenol p-eugenol p-allylguaiacol 4-allylcatechol-2-methyl ether 1,3,4-eugenol 4-allylguaiacol 丁香酚
Molecular Weight (g/mol): 164.2
Molecular Formula: C10H12O2
InChi Key: RRAFCDWBNXTKKO-UHFFFAOYSA-N
SMILES: COC1=CC(CC=C)=CC=C1O
Molecular Weight (g/mol) 
  • (5)
  • (6)
Quantity 
  • (1)
  • (3)
  • (1)
  • (2)
  • (2)
Boiling Point 
  • (2)
  • (4)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (4)
  • (2)
  • (3)
  • (2)
  • (5)
  • (6)
  • (1)
  • (3)
  • (1)
  • (2)
  • (2)
  • (2)
  • (4)
  • (4)
  • (1)
  • (1)
  • (1)
  • (1)