accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

NH2ONHNHOHNO

CAS RN 97-59-6

IUPAC Name: (2,5-dioxoimidazolidin-4-yl)urea
Synonyms: allantoin caswell no. 024 allantoin campbell brand woun'dres sebical 5-ureidohydrantoin allantoin (product) cutemol emollient HerpecinL glyoxylic diureide
Molecular Weight (g/mol): 158.12
Molecular Formula: C4H6N4O3
InChi Key: POJWUDADGALRAB-UHFFFAOYNA-N
SMILES: NC(=O)NC1NC(=O)NC1=O
Molecular Weight (g/mol) 
  • (3)
  • (7)
Percent Purity 
  • (3)
  • (5)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
Certifications/Compliance 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (3)
  • (3)
  • (7)
  • (3)
  • (5)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)