accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

H3COOCH3CH2

CAS RN 97-63-2

IUPAC Name: ethyl 2-methylprop-2-enoate
Synonyms: ethyl methacrylate
Molecular Weight (g/mol): 114.14
Molecular Formula: C6H10O2
InChi Key: SUPCQIBBMFXVTL-UHFFFAOYSA-N
SMILES: CCOC(=O)C(C)=C
Molecular Weight (g/mol) 
  • (4)
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
Boiling Point 
  • (2)
  • (2)
Percent Purity 
  • (2)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (2)
  • (2)
  • (2)
  • (4)
  • (3)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (1)