accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 97-65-4

HOOCH2OHO

CAS RN 97-65-4

IUPAC Name: 2-methylidenebutanedioic acid
Synonyms: itaconic acid propylenedicarboxylic acid ion(2-) methylenesuccinic acid
Molecular Weight (g/mol): 130.1
Molecular Formula: C5H6O4
InChi Key: LVHBHZANLOWSRM-UHFFFAOYSA-N
SMILES: OC(=O)CC(=C)C(O)=O
Molecular Weight (g/mol) 
  • (3)
  • (6)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
Percent Purity 
  • (3)
  • (3)
Physical Form 
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (3)
  • (3)
  • (1)
  • (3)
  • (6)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (3)
  • (3)
  • (3)
  • (3)