accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 97-67-6

IUPAC Name: (2S)-2-hydroxybutanedioic acid
Synonyms: (-)-malic acid S-2-hydroxybutanedioic acid malic acid L-(-)-form hydroxysuccinnic acid (-) S-(-)-malic acid S-(-)-malate L-apple acid S-2-hydroxybutanedioate
Molecular Weight (g/mol): 134.09
Molecular Formula: C4H6O5
InChi Key: BJEPYKJPYRNKOW-REOHCLBHSA-N
SMILES: O[C@@H](CC(O)=O)C(O)=O
Molecular Weight (g/mol) 
  • (15)
  • (3)
Quantity 
  • (2)
  • (1)
  • (5)
  • (1)
  • (3)
  • (1)
  • (5)
Percent Purity 
  • (3)
  • (2)
  • (3)
  • (7)
Physical Form 
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (4)
  • (3)
  • (3)
  • (15)
  • (3)
  • (2)
  • (1)
  • (5)
  • (1)
  • (3)
  • (1)
  • (5)
  • (3)
  • (2)
  • (3)
  • (7)
  • (3)