CAS RN 97-78-9
CAS RN 97-78-9
IUPAC Name:
2-(N-methyldodecanamido)acetic acid
Synonyms:
gardol
Molecular Weight (g/mol):
271.4
Molecular Formula:
C15H29NO3
InChi Key:
BACYUWVYYTXETD-UHFFFAOYSA-N
SMILES:
CCCCCCCCCCCC(=O)N(C)CC(O)=O
1
–
3
of
3
results
2
N-Lauroylsarcosine, 98.7%, MP Biomedicals™
CAS: 97-78-9 Molecular Formula: C15H29NO3 Molecular Weight (g/mol): 271.40 MDL Number: MFCD00021749 InChI Key: BACYUWVYYTXETD-UHFFFAOYSA-N Synonym: Hamposyl L PubChem CID: 7348 IUPAC Name: 2-(N-methyldodecanamido)acetic acid SMILES: CCCCCCCCCCCC(=O)N(C)CC(O)=O
3
N-Lauroylsarcosine, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.