accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 97096-16-7

IUPAC Name: 1,4-dioxaspiro[4.5]decan-8-amine
Synonyms: 1,4-dioxaspiro[4.5]decan-8-amine
Molecular Weight (g/mol): 157.21
Molecular Formula: C8H15NO2
InChi Key: KDAFVGCPLFJMHY-UHFFFAOYSA-N
SMILES: NC1CCC2(CC1)OCCO2

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results