accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CH3H3C(S)HNO(S)HONHO(S)NH2NNH2HNO(S)HNO(S)NCH3OONH(S)NH2ONH2O

CAS RN 97145-43-2

IUPAC Name: (2S)-2-[(2S)-2-[(2S)-2-[(2S)-2-[(2S)-2-{[(2S)-1-acetylpyrrolidin-2-yl]formamido}-3-phenylpropanamido]-5-[(diaminomethylidene)amino]pentanamido]-3-hydroxypropanamido]-3-methylbutanamido]pentanediamide
Synonyms: (2S)-2-[(2S)-2-[(2S)-2-[(2S)-2-[(2S)-2-{[(2S)-1-acetylpyrrolidin-2-yl]formamido}-3-phenylpropanamido]-5-[(diaminomethylidene)amino]pentanamido]-3-hydroxypropanamido]-3-methylbutanamido]pentanediamide
Molecular Weight (g/mol): 773.89
Molecular Formula: C35H55N11O9
InChi Key: OOZYLOMMGMLDHT-BIVGDOEESA-N
SMILES: CC(C)[C@H](NC(=O)[C@H](CO)NC(=O)[C@H](CCCN=C(N)N)NC(=O)[C@H](CC1=CC=CC=C1)NC(=O)[C@@H]1CCCN1C(C)=O)C(=O)N[C@@H](CCC(N)=O)C(N)=O
Product Type 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (2)