accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CH3OOHOOH3CNHNON

CAS RN 97161-97-2

IUPAC Name: methyl 16-hydroxy-15-methyl-3-oxo-28-oxa-4,14,19-triazaoctacyclo[12.11.2.1¹⁵,¹⁸.0²,⁶.0⁷,²⁷.0⁸,¹³.0¹⁹,²⁶.0²⁰,²⁵]octacosa-1(26),2(6),7(27),8(13),9,11,20,22,24-nonaene-16-carboxylate
Synonyms: staurosporinone
Molecular Weight (g/mol): 467.48
Molecular Formula: C27H21N3O5
InChi Key: KOZFSFOOLUUIGY-UHFFFAOYNA-N
SMILES: COC(=O)C1(O)CC2OC1(C)N1C3=C(C=CC=C3)C3=C1C1=C(C4=CC=CC=C4N21)C1=C3CNC1=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results