accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

H3COOOOH3COH3C

CAS RN 973-67-1

IUPAC Name: 5,6,7-trimethoxy-2-phenyl-4H-chromen-4-one
Synonyms: 5,6,7-trimethoxyflavone
Molecular Weight (g/mol): 312.32
Molecular Formula: C18H16O5
InChi Key: HJNJAUYFFFOFBW-UHFFFAOYSA-N
SMILES: COC1=C(OC)C(OC)=C2C(=O)C=C(OC2=C1)C1=CC=CC=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results