accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 975-17-7

HOOOHOOH

CAS RN 975-17-7

IUPAC Name: 2,6,7-trihydroxy-9-phenyl-3H-xanthen-3-one
Synonyms: 2,6,7-trihydroxy-9-phenylxanthen-3-one
Molecular Weight (g/mol): 320.3
Molecular Formula: C19H12O5
InChi Key: YDCFOUBAMGLLKA-UHFFFAOYSA-N
SMILES: OC1=CC2=C(C3=CC=CC=C3)C3=CC(O)=C(O)C=C3OC2=CC1=O
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results