accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

FNN

CAS RN 97509-75-6

IUPAC Name: 3-fluoropyridine-2-carbonitrile
Synonyms: 3-fluoropyridine-2-carbonitrile
Molecular Weight (g/mol): 122.1
Molecular Formula: C6H3FN2
InChi Key: VZFPSCNTFBJZHB-UHFFFAOYSA-N
SMILES: FC1=CC=CN=C1C#N
Molecular Weight (g/mol) 
  • (3)
  • (4)
Quantity 
  • (3)
  • (1)
  • (3)
Percent Purity 
  • (2)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (3)
  • (3)
  • (4)
  • (3)
  • (1)
  • (3)
  • (2)
  • (2)