accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

OOOOOFOO

CAS RN 97614-43-2

IUPAC Name: [3,5-bis(benzoyloxy)-4-fluorooxolan-2-yl]methyl benzoate
Synonyms: [3,5-bis(benzoyloxy)-4-fluorooxolan-2-yl]methyl benzoate
Molecular Weight (g/mol): 464.45
Molecular Formula: C26H21FO7
InChi Key: JOAHVPNLVYCSAN-UHFFFAOYNA-N
SMILES: FC1C(OC(=O)C2=CC=CC=C2)OC(COC(=O)C2=CC=CC=C2)C1OC(=O)C1=CC=CC=C1
Molecular Weight (g/mol) 
  • (3)
  • (2)
Quantity 
  • (1)
  • (1)
  • (1)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results