accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CH3OOOOOCH3CH3

CAS RN 97747-17-6

IUPAC Name: 2,2-dimethyl-2H,4H,4aH,8H,8aH-pyrano[3,2-d][1,3]dioxin-8-yl acetate
Synonyms: 2,2-dimethyl-4H,4aH,8H,8aH-pyrano[3,2-d][1,3]dioxin-8-yl acetate
Molecular Weight (g/mol): 228.24
Molecular Formula: C11H16O5
InChi Key: JSKNNUBJVJGXIN-UHFFFAOYNA-N
SMILES: CC(=O)OC1C=COC2COC(C)(C)OC12

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results