accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 978-86-9

OH

CAS RN 978-86-9

IUPAC Name: 4-(triphenylmethyl)phenol
Synonyms: 4-(triphenylmethyl)phenol
Molecular Weight (g/mol): 336.43
Molecular Formula: C25H20O
InChi Key: NIPKXTKKYSKEON-UHFFFAOYSA-N
SMILES: OC1=CC=C(C=C1)C(C1=CC=CC=C1)(C1=CC=CC=C1)C1=CC=CC=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results