accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 98-04-4

IUPAC Name: N,N,N-trimethylanilinium iodide
Synonyms: phenyl trimethyl ammonium iodide
Molecular Weight (g/mol): 263.12
Molecular Formula: C9H14IN
InChi Key: KKLAORVGAKUOPZ-UHFFFAOYSA-M
SMILES: [I-].C[N+](C)(C)C1=CC=CC=C1
Molecular Weight (g/mol) 
  • (1)
  • (3)
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (3)
  • (1)
  • (3)
  • (3)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)