accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 98-14-6

OHAsOHHOO

CAS RN 98-14-6

IUPAC Name: (4-hydroxyphenyl)arsonic acid
Synonyms: 4-hydroxyphenylarsonic acid
Molecular Weight (g/mol): 218.04
Molecular Formula: C6H7AsO4
InChi Key: RSRFURJMCNJVOW-UHFFFAOYSA-N
SMILES: OC1=CC=C(C=C1)[As](O)(O)=O
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results