accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 98-18-0

IUPAC Name: 3-aminobenzene-1-sulfonamide
Synonyms: M-aminobenzenesulfonamide
Molecular Weight (g/mol): 172.2
Molecular Formula: C6H8N2O2S
InChi Key: JPVKCHIPRSQDKL-UHFFFAOYSA-N
SMILES: NC1=CC=CC(=C1)S(N)(=O)=O
Percent Purity 
  • (2)
  • (3)
Quantity 
  • (1)
  • (2)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (3)
  • (2)
  • (3)
  • (1)
  • (2)
  • (1)
  • (1)