accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

ONOFFF

CAS RN 98-46-4

IUPAC Name: 1-nitro-3-(trifluoromethyl)benzene
Synonyms: M-trifluoromethylnitrobenzene
Molecular Weight (g/mol): 191.11
Molecular Formula: C7H4F3NO2
InChi Key: WHNAMGUAXHGCHH-UHFFFAOYSA-N
SMILES: [O-][N+](=O)C1=CC=CC(=C1)C(F)(F)F
Molecular Weight (g/mol) 
  • (2)
  • (1)
Boiling Point 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results