CAS RN 98-51-1
IUPAC Name:
1-tert-butyl-4-methylbenzene
Synonyms:
P-(T-butyl)toluene
Molecular Weight (g/mol):
148.25
Molecular Formula:
C11H16
InChi Key:
QCWXDVFBZVHKLV-UHFFFAOYSA-N
SMILES:
CC1=CC=C(C=C1)C(C)(C)C
1
–
2
of
2
results
1
4-tert-Butyltoluene, 96%
CAS: 98-51-1 Molecular Formula: C11H16 Molecular Weight (g/mol): 148.25 MDL Number: MFCD00008837 InChI Key: QCWXDVFBZVHKLV-UHFFFAOYSA-N Synonym: 4-tert-butyltoluene,p-tert-butyltoluene,1-tert-butyl-4-methylbenzene,p-t-butyltoluene,benzene, 1-1,1-dimethylethyl-4-methyl,4-t-butyltoluene,p-methyl-tert-butylbenzene,toluene, p-tert-butyl,1-methyl-4-tert-butylbenzene,4-methyl-tert-butylbenzene PubChem CID: 7390 IUPAC Name: 1-tert-butyl-4-methylbenzene SMILES: CC1=CC=C(C=C1)C(C)(C)C