accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

ClSOOI

CAS RN 98-61-3

IUPAC Name: 4-iodobenzene-1-sulfonyl chloride
Synonyms: 4-iodobenzenesulfonyl chloride
Molecular Weight (g/mol): 302.51
Molecular Formula: C6H4ClIO2S
InChi Key: POXFXTSTVWDWIR-UHFFFAOYSA-N
SMILES: ClS(=O)(=O)C1=CC=C(I)C=C1
Quantity 
  • (1)
  • (2)
  • (2)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (3)
  • (1)
  • (2)
  • (2)
  • (2)